Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258024.1 | NP_001244953.1 | 11 | 2262 | ||
XM_017022173.1 | XP_016877662.1 | 5 | 2675 | |||
GenBank mRNA | AY669507.1 | - | 11 | 2262 |