Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC014514.1 | - | 1, 2 | 148 | 3542845 | |
BT007361.1 | - | 1, 2 | 78 |