Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278595.1 | NP_001265524.1 | 12 | 2502 | ||
NM_005916.4 | NP_005907.3 | 13 | 2427 | |||
NM_182776.2 | NP_877577.1 | 12 | 2393 | |||
XM_005250348.3 | XP_005250405.1 | 13 | 2081 | |||
XM_017012217.1 | XP_016867706.1 | 13 | 1958 | |||
GenBank mRNA | AF279900.1 | - | 4 | 471 | ||
AK055379.1 | - | 12 | 2370 | |||
AK096959.1 | - | 14 | 1791 | |||
AK226175.1 | - | 12 | 1932 | |||
AK293172.1 | - | 13 | 1829 | |||
AK314575.1 | - | 13 | 1894 | |||
AY007130.1 | - | 5 | 724 | |||
BC013375.2 | - | 13 | 1863 | 3946342 | ||
D55716.1 | - | 13 | 1897 | |||
D86748.1 | - | 13 | 1867 | |||
KU178172.1 | - | 13 | 1781 | |||
KU178173.1 | - | 12 | 1856 | |||
X74796.1 | - | 10 | 1477 |