Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282403.1 | NP_001269332.1 | 7, 8 | 908 | ||
NM_001282404.1 | NP_001269333.1 | 7, 8 | 865 | |||
NM_005918.3 | NP_005909.2 | 8, 9 | 1034 | |||
GenBank mRNA | AF047470.1 | - | 8, 9 | 978 | ||
AK290779.1 | - | 8, 9 | 968 | |||
AK316587.1 | - | 8, 9 | 917 | |||
BC001917.1 | - | 8, 9 | 920 | 2823443 | ||
CR536548.1 | - | 8, 9 | 892 | |||
DQ402957.1 | - | 7, 8 | 829 |