Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145337.2 | NP_001138809.1 | 2, 3 | 156 | ||
NM_001145339.2 | NP_001138811.1 | 2, 3 | 406 | |||
NM_001145340.2 | NP_001138812.1 | 2, 3 | 156 | |||
NM_001278462.1 | NP_001265391.1 | 2, 3 | 156 | |||
NM_002392.5 | NP_002383.2 | 2, 3 | 406 | |||
XM_005268872.4 | XP_005268929.1 | 2, 3 | 334 | |||
XM_006719400.3 | XP_006719463.1 | 2, 3 | 429 | |||
GenBank mRNA | AF092843.1 | - | 2, 3 | 114 | ||
AF092844.1 | - | 2, 3 | 114 | |||
AF092845.1 | - | 2, 3 | 114 | |||
AF385322.1 | - | 1, 2 | 82 | |||
AF385323.1 | - | 1, 2 | 82 | |||
AF385324.1 | - | 1, 2 | 82 | |||
AF385326.1 | - | 1, 2 | 82 | |||
AF385327.1 | - | 1, 2 | 82 | |||
AJ278975.1 | - | 1, 2 | 82 | |||
AJ278977.1 | - | 1, 2 | 82 | |||
AJ430612.1 | - | 1, 2 | 82 | |||
AJ430614.1 | - | 1, 2 | 82 | |||
AJ491698.1 | - | 1, 2 | 82 | |||
AJ491699.1 | - | 1, 2 | 82 | |||
AJ491700.1 | - | 1, 2 | 82 | |||
AJ491701.1 | - | 1, 2 | 82 | |||
AJ550517.1 | - | 1, 2 | 82 | |||
AJ550518.1 | - | 1, 2 | 82 | |||
AJ550519.1 | - | 1, 2 | 82 | |||
AJ550520.1 | - | 1, 2 | 82 | |||
AK290341.1 | - | 2, 3 | 356 | |||
AM902642.1 | - | 1, 2 | 82 | |||
AM902643.1 | - | 1, 2 | 82 | |||
AM902644.1 | - | 1, 2 | 82 | |||
AM902646.1 | - | 1, 2 | 82 | |||
AM902647.1 | - | 1, 2 | 82 | |||
AM902648.1 | - | 1, 2 | 82 | |||
AM902649.1 | - | 1, 2 | 82 | |||
AM902650.1 | - | 1, 2 | 82 | |||
AM902651.1 | - | 1, 2 | 82 | |||
AM902654.1 | - | 1, 2 | 82 | |||
AM902655.1 | - | 1, 2 | 82 | |||
AM902656.1 | - | 1, 2 | 82 | |||
AM902657.1 | - | 1, 2 | 82 | |||
AM902658.1 | - | 1, 2 | 82 | |||
AM902659.1 | - | 1, 2 | 82 | |||
AM902660.1 | - | 1, 2 | 82 | |||
AM902661.1 | - | 1, 2 | 82 | |||
AM902663.1 | - | 1, 2 | 82 | |||
AM902664.1 | - | 1, 2 | 82 | |||
AM902665.1 | - | 1, 2 | 82 | |||
AM902666.1 | - | 1, 2 | 82 | |||
AM902667.1 | - | 1, 2 | 82 | |||
AM902668.1 | - | 1, 2 | 82 | |||
AM902669.1 | - | 1, 2 | 82 | |||
AM902672.1 | - | 1, 2 | 82 | |||
AM902673.1 | - | 1, 2 | 82 | |||
AM902674.1 | - | 1, 2 | 82 | |||
AM902675.1 | - | 1, 2 | 82 | |||
AM902676.1 | - | 1, 2 | 82 | |||
AM902678.1 | - | 1, 2 | 82 | |||
AM902679.1 | - | 1, 2 | 82 | |||
AM902680.1 | - | 1, 2 | 82 | |||
AM902681.1 | - | 1, 2 | 82 | |||
BC152384.1 | - | 1, 2 | 85 | |||
BC152390.1 | - | 1, 2 | 85 | |||
BE296905.1 | - | 2, 3 | 371 | |||
BT007258.1 | - | 1, 2 | 82 | |||
EU076746.1 | - | 2, 3 | 156 | |||
EU076748.1 | - | 2, 3 | 156 | |||
EU076749.1 | - | 2, 3 | 156 | |||
HM213901.1 | - | 1, 2 | 82 | |||
HM213902.1 | - | 1, 2 | 82 | |||
HM213903.1 | - | 1, 2 | 82 | |||
HM213904.1 | - | 1, 2 | 82 | |||
HY174841.1 | - | 2, 3 | 408 | |||
JX015379.1 | - | 1, 2 | 82 | |||
JX015380.1 | - | 1, 2 | 82 | |||
JX015381.1 | - | 1, 2 | 82 | |||
JX015382.1 | - | 1, 2 | 82 | |||
JX015383.1 | - | 1, 2 | 82 | |||
JX015384.1 | - | 1, 2 | 82 | |||
JX015385.1 | - | 1, 2 | 82 | |||
JX015387.1 | - | 1, 2 | 82 | |||
JX015388.1 | - | 1, 2 | 82 | |||
JX015389.1 | - | 1, 2 | 82 | |||
JX015390.1 | - | 1, 2 | 82 | |||
JX015391.1 | - | 1, 2 | 82 | |||
JX015392.1 | - | 1, 2 | 82 | |||
M92424.1 | - | 2, 3 | 393 | |||
U33201.1 | - | 1, 2 | 82 | |||
U33203.1 | - | 1, 2 | 82 | |||
Z12020.1 | - | 2, 3 | 393 |