Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000249.3 | NP_000240.1 | 9 | 894 | ||
NM_001167617.1 | NP_001161089.1 | 9 | 689 | |||
NM_001167618.1 | NP_001161090.1 | 9 | 689 | |||
NM_001167619.1 | NP_001161091.1 | 8 | 602 | |||
NM_001258271.1 | NP_001245200.1 | 9 | 894 | |||
NM_001258273.1 | NP_001245202.1 | 8 | 803 | |||
NM_001258274.1 | NP_001245203.1 | 10 | 839 | |||
XM_005265163.1 | XP_005265220.1 | 9 | 694 | |||
XM_005265164.1 | XP_005265221.1 | 9 | 747 | |||
XM_005265166.1 | XP_005265223.1 | 9 | 702 | |||
XM_011533727.1 | XP_011532029.1 | 9 | 702 | |||
XM_017006451.1 | XP_016861940.1 | 9 | 747 | |||
GenBank mRNA | AB209848.1 | - | 9 | 672 | ||
AK222810.1 | - | 9 | 701 | |||
AK295359.1 | - | 9 | 668 | |||
AK298324.1 | - | 9 | 640 | |||
AK298583.1 | - | 9 | 662 | |||
AK302807.1 | - | 11 | 623 | |||
AK311365.1 | - | 9 | 643 | |||
AK312609.1 | - | 9 | 710 | |||
AK316074.1 | - | 8 | 575 | |||
AK316264.1 | - | 8 | 620 | |||
BC006850.1 | - | 9 | 746 | 3451538 | ||
BX648844.1 | - | 10 | 843 | |||
DQ648890.1 | - | 2 | 77 | |||
EU188667.1 | - | 2 | 37 | |||
EU188669.1 | - | 2 | 37 | |||
EU188670.1 | - | 2 | 37 | |||
EU188671.1 | - | 1 | 36 | |||
U07343.1 | - | 9 | 717 | |||
U07418.1 | - | 9 | 737 |