Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004531.4 | NP_004522.1 | 2 | 548 | ||
NM_176806.3 | NP_789776.1 | 2 | 87 | |||
GenBank mRNA | AF091871.1 | - | 2 | 60 | ||
AF117815.1 | - | 2 | 71 | |||
AF155659.1 | - | 2 | 273 | |||
AK312887.1 | - | 2 | 72 | |||
BC029287.1 | - | 2 | 50 | |||
BC039720.2 | - | 1 | 28 | |||
BC046097.1 | - | 2 | 234 | 5740329 | ||
BC095417.1 | - | 2 | 88 | |||
CD514049.1 | - | 2 | 83 | |||
DA967651.1 | - | 2 | 531 |