Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG028932.1 | - | 31 |
Gene Symbol: RRAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102669.2 | NP_001096139.1 | 2 | 301 | ||
NM_001177314.1 | NP_001170785.1 | 2 | 146 | |||
NM_001177315.1 | NP_001170786.1 | 2 | 278 | |||
NM_012250.5 | NP_036382.2 | 2 | 452 | |||
XM_017017363.1 | XP_016872852.1 | 2 | 117 | |||
GenBank mRNA | AF493924.1 | - | 2 | 135 | ||
AK299606.1 | - | 3 | 302 | |||
AK300103.1 | - | 2 | 146 | |||
AK302033.1 | - | 2 | 118 | |||
AK309550.1 | - | 2 | 278 | |||
AK309683.1 | - | 2 | 259 | |||
AK313976.1 | - | 2 | 451 | |||
BC013106.1 | - | 2 | 335 | 4389214 | ||
BG428983.1 | - | 2 | 301 | |||
BP206021.1 | - | 2 | 451 | |||
BX324335.2 | - | 2 | 438 | |||
CB961515.1 | - | 439 | ||||
DC368033.1 | - | 2 | 259 | |||
M31468.1 | - | 2 | 135 |