Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CITED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB451286.1 | - | 2 | 298 | ||
AB451418.1 | - | 2 | 298 | |||
BC004240.1 | - | 3 | 827 | 3627230 | ||
BP247232.1 | - | 3 | 453 | |||
U65092.1 | - | 3 | 493 |