Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102669.2 | NP_001096139.1 | 6 | 733 | ||
NM_001177314.1 | NP_001170785.1 | 6 | 578 | |||
NM_001177315.1 | NP_001170786.1 | 6 | 710 | |||
NM_012250.5 | NP_036382.2 | 6 | 884 | |||
XM_017017363.1 | XP_016872852.1 | 6 | 549 | |||
GenBank mRNA | AF493924.1 | - | 6 | 567 | ||
AK299606.1 | - | 7 | 734 | |||
AK300103.1 | - | 6 | 578 | |||
AK302033.1 | - | 6 | 550 | |||
AK309550.1 | - | 6 | 710 | |||
AK309683.1 | - | 6 | 691 | |||
AK313976.1 | - | 6 | 883 | |||
BC013106.1 | - | 6 | 767 | 4389214 | ||
M31468.1 | - | 6 | 567 |
Gene Symbol: SCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG028932.1 | - | 460 |