Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005592.3 | NP_005583.1 | 9 | 1204 | ||
XM_005251994.3 | XP_005252051.1 | 10 | 1816 | |||
XM_005251995.3 | XP_005252052.1 | 10 | 1815 | |||
XM_005251996.3 | XP_005252053.1 | 9 | 1784 | |||
GenBank mRNA | AF006464.1 | - | 9 | 1116 |