Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MVK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000431.3 | NP_000422.1 | 8, 9 | 959 | ||
NM_001114185.2 | NP_001107657.1 | 8, 9 | 950 | |||
NM_001301182.1 | NP_001288111.1 | 7, 8 | 803 | |||
XM_011538372.2 | XP_011536674.2 | 8, 9 | 1067 | |||
XM_017019313.1 | XP_016874802.1 | 7, 8 | 756 | |||
XM_017019314.1 | XP_016874803.1 | 8, 9 | 1425 | |||
GenBank mRNA | AB209722.1 | - | 8, 9 | 842 | ||
AK023087.1 | - | 1, 2 | 1295 | |||
AK293130.1 | - | 6, 7 | 526 | |||
AK295338.1 | - | 7, 8 | 661 | |||
AK315678.1 | - | 8, 9 | 838 | |||
BC016140.1 | - | 8, 9 | 802 | 3921543 | ||
KU178205.1 | - | 7, 8 | 766 | |||
KU178206.1 | - | 6, 7 | 610 | |||
M88468.1 | - | 8, 9 | 857 |