Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NQO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000904.4 | NP_000895.2 | 4 | 579 | ||
NM_001290221.1 | NP_001277150.1 | 7 | 1094 | |||
NM_001290222.1 | NP_001277151.1 | 4 | 579 | |||
NM_001318940.1 | NP_001305869.1 | 4 | 519 | |||
GenBank mRNA | AK056981.1 | - | 7 | 936 | ||
AK311746.1 | - | 4 | 538 | |||
BC006096.2 | - | 3 | 310 | 3507945 | ||
BI333292.1 | - | 4 | 518 | |||
BI670476.1 | - | 4 | 567 | |||
CD014013.1 | - | 4 | 531 | |||
J02888.1 | - | 4 | 399 |