Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001680.4 | NP_001671.2 | 6 | 479 | ||
NM_021603.3 | NP_067614.1 | 6 | 486 | |||
GenBank mRNA | AF241236.1 | - | 6 | 454 | ||
BC005302.1 | - | 6 | 486 | 3828677 | ||
BC013289.1 | - | 6 | 482 | 3686762 | ||
BX641053.1 | - | 1 | 2109 | |||
U50743.1 | - | 6 | 443 |
Gene Symbol: FXYD6-FXYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204268.1 | NP_001191197.1 | 11 | 1092 | ||
NM_001243598.2 | NP_001230527.1 | 10 | 1030 |