Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317960.1 | NP_001304889.1 | 5, 6 | 770 | ||
NM_012255.4 | NP_036387.2 | 5, 6 | 610 | |||
XM_017027722.1 | XP_016883211.1 | 5, 6 | 765 | |||
XM_017027723.1 | XP_016883212.1 | 5, 6 | 539 | |||
XM_017027724.1 | XP_016883213.1 | 5, 6 | 765 | |||
GenBank mRNA | AF064257.1 | - | 5, 6 | 563 | ||
AF152169.1 | - | 5, 6 | 513 | |||
AK001573.1 | - | 5, 6 | 580 | |||
AK302846.1 | - | 5, 6 | 474 | |||
AK303312.1 | - | 4, 5 | 421 | |||
AK314602.1 | - | 5, 6 | 549 | |||
AL136841.1 | - | 5, 6 | 536 | |||
AY382900.1 | - | 4, 5 | 425 | |||
CF994999.1 | - | 5, 6 | 617 |