Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000436.3 | NP_000427.1 | 8 | 1142 | ||
XM_017009494.1 | XP_016864983.1 | 8 | 1023 | |||
GenBank mRNA | AK299668.1 | - | 4 | 330 | ||
AK312327.1 | - | 8 | 838 | |||
BC009001.1 | - | 8 | 866 | 4148643 | ||
CR457049.1 | - | 8 | 811 | |||
U62961.1 | - | 8 | 909 |