Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015713.4 | NP_056528.2 | 1, 2 | 294 | ||
GenBank mRNA | AB036063.1 | - | 1, 2 | 294 | ||
AB163438.1 | - | 1, 2 | 50 | |||
AB166671.1 | - | 1, 2 | 50 | |||
AK001965.1 | - | 1, 2 | 89 | |||
AL137348.1 | - | 1, 2 | 96 | |||
BC108261.1 | - | 1, 2 | 106 | 6143754 | ||
BC117496.1 | - | 1, 2 | 131 | 40126047 | ||
BC130628.1 | - | 1, 2 | 131 | 40146659 |