Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172477.1 | NP_001165948.1 | 3 | 529 | ||
NM_001172478.1 | NP_001165949.1 | 2 | 392 | |||
NM_015713.4 | NP_056528.2 | 3 | 548 | |||
GenBank mRNA | AB036063.1 | - | 3 | 548 | ||
AB163437.1 | - | 2 | 148 | |||
AB166671.1 | - | 3 | 304 | |||
AK001965.1 | - | 3 | 343 | |||
AK304354.1 | - | 3 | 473 | |||
AL137348.1 | - | 3 | 350 | |||
BC042468.1 | - | 1 | 106 | |||
BC108261.1 | - | 3 | 360 | 6143754 | ||
BC117496.1 | - | 3 | 385 | 40126047 | ||
BC130628.1 | - | 3 | 385 | 40146659 | ||
CR627376.1 | - | 3 | 473 | |||
DC308409.1 | - | 3 | 529 |