Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IKZF2 SIRNA Assays ›
Gene Symbol:
IKZF2
Gene Name:
IKAROS family zinc finger 2
Gene Aliases:
ANF1A2, HELIOS, ZNF1A2, ZNFN1A2
Chromosome Location:
Chr.2: 212999684 - 213152753 on Build GRCh38
UniGene ID:
Hs.604950
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22418
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IKZF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079526.1
NP_001072994.1
8
990
NM_016260.2
NP_057344.2
7
921
XM_005246384.4
XP_005246441.1
7
1031
XM_005246385.3
XP_005246442.1
7
1061
XM_005246386.4
XP_005246443.1
7
912
XM_011510802.2
XP_011509104.1
8
1103
XM_011510803.2
XP_011509105.1
8
952
XM_011510804.2
XP_011509106.1
8
875
XM_011510805.2
XP_011509107.1
8
964
XM_011510807.2
XP_011509109.1
8
1139
XM_011510808.2
XP_011509110.1
8
989
XM_011510809.2
XP_011509111.1
8
1001
XM_011510810.2
XP_011509112.1
8
915
XM_011510811.2
XP_011509113.1
7
1036
XM_011510812.2
XP_011509114.1
7
897
XM_011510814.2
XP_011509116.1
7
1395
XM_011510815.2
XP_011509117.1
7
843
XM_011510816.2
XP_011509118.1
5
844
XM_011510818.2
XP_011509120.1
5
6498
XM_017003589.1
XP_016859078.1
7
983
XM_017003590.1
XP_016859079.1
7
953
XM_017003591.1
XP_016859080.1
5
1386
GenBank mRNA
AF130863.1
-
7
921
AK301503.1
-
7
917
AY008294.1
-
1
1385
AY587062.1
-
8
905
AY587063.1
-
8
867
AY587064.1
-
8
951
AY587065.1
-
7
758
AY587067.1
-
6
569
AY587068.1
-
7
829
AY587069.1
-
6
674
AY587070.1
-
4
317
AY587071.1
-
5
536
AY587073.1
-
7
931
BC028936.1
-
8
990
4540576
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
biological_process
positive regulation of transcription from RNA polymerase II promoter
molecular_function
transcription factor activity, sequence-specific DNA binding
protein binding
protein homodimerization activity
transcription regulatory region DNA binding
metal ion binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline