Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113546.1 | NP_001107018.1 | 4 | 769 | ||
NM_001113547.1 | NP_001107019.1 | 1 | 650 | |||
NM_016357.4 | NP_057441.1 | 4 | 769 | |||
XM_011538455.2 | XP_011536757.1 | 4 | 829 | |||
XM_017019430.1 | XP_016874919.1 | 4 | 683 | |||
GenBank mRNA | AB209512.1 | - | 4 | 705 | ||
AF198454.1 | - | 4 | 696 | |||
AF198455.1 | - | 1 | 588 | |||
AK000335.1 | - | 1 | 293 | |||
AK000372.1 | - | 4 | 679 | |||
AK222971.1 | - | 4 | 692 | |||
AK297766.1 | - | 2 | 365 | |||
AK314447.1 | - | 4 | 681 | |||
AL136911.1 | - | 4 | 688 | |||
BC001247.1 | - | 1 | 301 | |||
BC110815.1 | - | 4 | 706 | 5115521 | ||
BC136763.1 | - | 4 | 655 | 9020753 | ||
BC144402.1 | - | 4 | 655 | |||
BC144403.1 | - | 4 | 655 | |||
DA738195.1 | - | 1 | 650 |