Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016577.3 | NP_057661.3 | 7 | 906 | ||
XM_011512893.2 | XP_011511195.1 | 7 | 581 | |||
GenBank mRNA | AB073348.1 | - | 1 | 25 | ||
AF166492.1 | - | 7 | 862 | |||
AF498940.1 | - | 7 | 525 | |||
AK295451.1 | - | 8 | 643 | |||
AK312378.1 | - | 7 | 826 | |||
BC002510.2 | - | 7 | 875 | 3050592 | ||
BC078662.1 | - | 7 | 654 | 6450986 | ||
BT007263.1 | - | 7 | 525 |