Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015983.3 | NP_057067.1 | 7 | 1182 | ||
XM_006715734.3 | XP_006715797.1 | 8 | 769 | |||
XM_011515420.2 | XP_011513722.1 | 8 | 1275 | |||
XM_011515422.2 | XP_011513724.1 | 6 | 1119 | |||
XM_011515423.2 | XP_011513725.1 | 5 | 553 | |||
XM_017012292.1 | XP_016867781.1 | 9 | 862 | |||
XM_017012295.1 | XP_016867784.1 | 6 | 616 | |||
GenBank mRNA | AF125044.1 | - | 7 | 1221 | ||
AF125045.1 | - | 6 | 1068 | |||
AK001446.1 | - | 7 | 1170 | |||
BC004104.2 | - | 7 | 1174 | 3835609 |