Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017420.4 | NP_059116.3 | 3 | 2220 | ||
XM_005267759.2 | XP_005267816.1 | 4 | 2199 | |||
GenBank mRNA | AK308528.1 | - | 4 | 1577 | ||
BC098135.1 | - | 3 | 2097 | 40007527 | ||
BC098282.1 | - | 3 | 2097 | 40007529 | ||
BC099722.1 | - | 3 | 2097 | 40007530 | ||
BC101934.1 | - | 3 | 2097 | 40007526 |