Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304451.1 | NP_001291380.1 | 8 | 1001 | ||
NM_001304452.1 | NP_001291381.1 | 9 | 1146 | |||
NM_002631.3 | NP_002622.2 | 9 | 1067 | |||
GenBank mRNA | AK290404.1 | - | 9 | 992 | ||
AK296889.1 | - | 10 | 895 | |||
AK298830.1 | - | 9 | 1146 | |||
AK300952.1 | - | 8 | 904 | |||
AK303898.1 | - | 7 | 654 | |||
AK304423.1 | - | 8 | 924 | |||
BC000368.2 | - | 9 | 978 | 2819330 | ||
U30255.1 | - | 8 | 947 |