Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193646.1 | NP_001180575.1 | 3 | 1160 | ||
NM_001290746.1 | NP_001277675.1 | 3 | 1131 | |||
NM_012068.5 | NP_036200.2 | 4 | 1365 | |||
XM_011526629.2 | XP_011524931.1 | 3 | 1067 | |||
GenBank mRNA | AB021663.2 | - | 3 | 1102 | ||
AB073613.1 | - | 3 | 1098 | |||
AF101388.1 | - | 303 | ||||
AF305687.1 | - | 2 | 833 | |||
AK024402.1 | - | 3 | 1102 | |||
AK091985.1 | - | 2 | 1074 | |||
AK290592.1 | - | 4 | 1304 | |||
BC005174.2 | - | 3 | 1089 | 3504701 |