Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002641.3 | NP_002632.1 | 5 | 1260 | ||
NM_020473.3 | NP_065206.3 | 4 | 483 | |||
NR_033835.1 | 5 | 886 | ||||
NR_033836.1 | 5 | 602 | ||||
XM_011545539.2 | XP_011543841.1 | 4 | 2084 | |||
XM_017029581.1 | XP_016885070.1 | 4 | 1207 | |||
GenBank mRNA | AK098047.1 | - | 4 | 696 | ||
AK303538.1 | - | 4 | 475 | |||
AK304144.1 | - | 4 | 1377 | |||
BC038236.1 | - | 4 | 1217 | 5733899 | ||
CX870731.1 | - | 4 | 562 | |||
D11466.1 | - | 5 | 1229 | |||
S74936.1 | - | 3 | 702 |