Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI4KA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058004.3 | NP_477352.3 | 39 | 4710 | ||
XM_005261634.1 | XP_005261691.1 | 38 | 4617 | |||
XM_017028829.1 | XP_016884318.1 | 38 | 4644 | |||
XM_017028830.1 | XP_016884319.1 | 25 | 2866 | |||
GenBank mRNA | AB210002.1 | - | 39 | 4680 | ||
AF012872.1 | - | 38 | 4449 | |||
AK302424.1 | - | 39 | 4638 | |||
AK314781.1 | - | 11 | 1364 | |||
BC018120.1 | - | 9 | 1027 | |||
BC053654.1 | - | 8 | 1003 | |||
L36151.1 | - | 7 | 996 |