Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278717.1 | NP_001265646.1 | 5 | 427 | ||
NM_001278718.1 | NP_001265647.1 | 5 | 321 | |||
NM_005031.4 | NP_005022.2 | 5 | 317 | |||
NM_021902.3 | NP_068702.1 | 5 | 293 | |||
XM_017026874.1 | XP_016882363.1 | 5 | 272 | |||
XM_017026875.1 | XP_016882364.1 | 5 | 487 | |||
XM_017026876.1 | XP_016882365.1 | 5 | 422 | |||
GenBank mRNA | AK289811.1 | - | 5 | 260 | ||
BC032800.2 | - | 4 | 187 | 5179163 | ||
BM691130.1 | - | 5 | 260 | |||
BM856555.1 | - | 5 | 254 | |||
BX115036.1 | - | 5 | 317 | |||
GQ258667.1 | - | 4 | 165 | |||
H57207.1 | - | 5 | 318 | |||
HY107209.1 | - | 5 | 294 | |||
HY111815.1 | - | 5 | 321 | |||
HY151101.1 | - | 5 | 282 | |||
HY151827.1 | - | 5 | 429 | |||
U72245.1 | - | 4 | 183 |