Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136007.1 | NP_001129479.1 | 11 | 1486 | ||
NM_001136011.1 | NP_001129483.1 | 10 | 1364 | |||
NM_001136012.1 | NP_001129484.1 | 8 | 1358 | |||
NM_005971.3 | NP_005962.1 | 9 | 1280 | |||
NM_021910.2 | NP_068710.1 | 9 | 1442 | |||
GenBank mRNA | BC090044.1 | - | 9 | 1229 | 30395606 | |
U28249.1 | - | 9 | 1368 | |||
U31214.1 | - | 1 | 357 |