Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316323.1 | NP_001303252.1 | 6 | 485 | ||
NM_001316324.1 | NP_001303253.1 | 7 | 633 | |||
NM_002695.3 | NP_002686.2 | 7 | 660 | |||
XM_011528070.2 | XP_011526372.1 | 7 | 658 | |||
XM_017026883.1 | XP_016882372.1 | 6 | 477 | |||
GenBank mRNA | AK122813.1 | - | 6 | 1822 | ||
AK295975.1 | - | 7 | 578 | |||
AK312625.1 | - | 7 | 613 | |||
BC004441.1 | - | 7 | 605 | 2820565 | ||
BC034144.1 | - | 7 | 625 | 4617000 | ||
D38251.1 | - | 7 | 613 | |||
DC351659.1 | - | 6 | 468 | |||
J04965.1 | - | 7 | 607 | |||
S42643.1 | - | 2 | 97 |