Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT1A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019075.2 | NP_061948.1 | 1 | 799 | ||
GenBank mRNA | AK025403.1 | - | 1 | 406 | ||
AY435137.1 | - | 1 | 753 | |||
BC020971.1 | - | 1 | 822 | 3850654 | ||
BC053576.1 | - | 1 | 774 | 4452440 | ||
BC069210.1 | - | 1 | 783 | 4660540 | ||
U89508.1 | - | 1 | 753 |
Gene Symbol: UGT1A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019077.2 | NP_061950.2 | 1 | 753 | ||
GenBank mRNA | AY435142.1 | - | 1 | 753 | ||
BC001521.2 | - | 1 | 323 | |||
DQ383513.1 | - | 1 | 465 | |||
U89507.1 | - | 1 | 753 |
Gene Symbol: UGT1A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019076.4 | NP_061949.3 | 1 | 816 | ||
GenBank mRNA | AF030310.1 | - | 1 | 753 | ||
AF462267.1 | - | 1 | 786 | |||
AF462268.1 | - | 1 | 816 | |||
AK313488.1 | - | 1 | 799 | |||
AY435143.1 | - | 1 | 753 | |||
DQ364251.1 | - | 1 | 408 |
Gene Symbol: UGT1A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021027.2 | NP_066307.1 | 1 | 790 | ||
GenBank mRNA | AF056188.1 | - | 1 | 771 | ||
AK313510.1 | - | 1 | 829 | |||
AY435144.1 | - | 1 | 753 | |||
BC058844.1 | - | 1 | 796 | 4593613 | ||
DQ364246.1 | - | 1 | 105 | |||
S55985.1 | - | 1 | 787 |