Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017573.4 | NP_060043.2 | 6, 7 | 786 | ||
XM_011528085.2 | XP_011526387.1 | 7, 8 | 1011 | |||
XM_011528086.2 | XP_011526388.1 | 7, 8 | 1011 | |||
XM_011528087.2 | XP_011526389.1 | 7, 8 | 1011 | |||
XM_011528088.2 | XP_011526390.1 | 7, 8 | 663 | |||
XM_011528091.2 | XP_011526393.1 | 5, 6 | 484 | |||
XM_011528092.2 | XP_011526394.1 | 7, 8 | 1011 | |||
XM_011528093.2 | XP_011526395.1 | 7, 8 | 1011 | |||
GenBank mRNA | AY358963.1 | - | 6, 7 | 744 |