Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYTL2 SIRNA Assays ›
Gene Symbol:
SYTL2
Gene Name:
synaptotagmin like 2
Gene Aliases:
CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A
Chromosome Location:
Chr.11: 85694221 - 85811182 on Build GRCh38
UniGene ID:
Hs.369520
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s224321
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYTL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001162951.2
NP_001156423.1
13, 14
2444
NM_001162952.2
NP_001156424.1
5, 6
974
NM_001162953.2
NP_001156425.1
13, 14
2447
NM_001289608.1
NP_001276537.1
14, 15
2253
NM_001289609.1
NP_001276538.1
6, 7
1087
NM_001289610.1
NP_001276539.1
6, 7
658
NM_032943.4
NP_116561.1
14, 15
2623
NM_206929.3
NP_996812.1
7, 8
778
NM_206930.3
NP_996813.1
4, 5
951
XM_005274057.1
XP_005274114.1
13, 14
6083
XM_005274060.1
XP_005274117.1
14, 15
2267
XM_005274061.1
XP_005274118.1
14, 15
2264
XM_005274062.2
XP_005274119.1
15, 16
2688
XM_005274063.1
XP_005274120.1
13, 14
2147
XM_005274066.1
XP_005274123.1
12, 13
2096
XM_005274067.4
XP_005274124.1
6, 7
2390
XM_011545105.1
XP_011543407.1
14, 15
6131
XM_011545106.1
XP_011543408.1
14, 15
6128
XM_011545107.2
XP_011543409.1
15, 16
6552
XM_011545108.1
XP_011543410.1
13, 14
6011
XM_011545109.2
XP_011543411.1
14, 15
6113
XM_011545114.2
XP_011543416.1
5, 6
2296
XM_011545115.1
XP_011543417.1
3, 4
390
XM_011545116.2
XP_011543418.1
4, 5
2175
XM_017017933.1
XP_016873422.1
13, 14
6080
XM_017017934.1
XP_016873423.1
15, 16
6549
XM_017017935.1
XP_016873424.1
11, 12
5927
XM_017017936.1
XP_016873425.1
11, 12
5625
XM_017017937.1
XP_016873426.1
14, 15
2168
XM_017017938.1
XP_016873427.1
13, 14
2126
XM_017017939.1
XP_016873428.1
7, 8
2434
XM_017017940.1
XP_016873429.1
6, 7
2386
GenBank mRNA
AB046817.1
-
14, 15
2634
AJ303364.1
-
2, 3
270
AK024872.1
-
4, 5
1115
AK074737.1
-
7, 8
735
AK092121.1
-
4, 5
769
AK124754.1
-
7, 8
2359
AK127504.1
-
8, 9
3322
AK131365.1
-
7, 8
1251
AK222707.1
-
4, 5
1115
AK222710.1
-
4, 5
1115
AK296224.1
-
14, 15
2235
AK296740.1
-
11, 12
2183
AK298604.1
-
6, 7
576
AL601023.1
-
6, 7
631
AL834422.1
-
1, 2
13
AY386362.1
-
7, 8
4638
BC015540.2
-
4, 5
445
BC068495.1
-
6, 7
812
BC110315.1
-
5, 6
503
BC136450.1
-
13, 14
2379
9020439
BC144114.1
-
13, 14
2379
9052634
BC144115.1
-
13, 14
2376
9052635
BX647671.1
-
8, 9
3074
BX648068.1
-
7, 8
2520
EU035829.1
-
13, 14
2100
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
exocytosis
vesicle docking involved in exocytosis
vesicle fusion
negative regulation of phosphatase activity
vesicle-mediated transport
regulation of calcium ion-dependent exocytosis
calcium ion-regulated exocytosis of neurotransmitter
positive regulation of mucus secretion
protein localization to plasma membrane
phosphatidylserine binding
calcium ion binding
protein binding
calcium-dependent phospholipid binding
phosphatidylinositol-4,5-bisphosphate binding
Rab GTPase binding
phosphatase binding
syntaxin binding
clathrin binding
neurexin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline