Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177983.2 | NP_817092.1 | 7 | 1386 | ||
GenBank mRNA | AB209197.1 | - | 5 | 2832 | ||
AK127593.1 | - | 7 | 1252 | |||
AK293141.1 | - | 8 | 1180 | |||
AK312455.1 | - | 7 | 1376 | |||
BC000057.2 | - | 7 | 1235 | 3505271 | ||
BC007361.2 | - | 3 | 487 | |||
BC022061.2 | - | 7 | 1232 | 2988852 | ||
BT007454.1 | - | 7 | 1125 | |||
CR457059.1 | - | 7 | 1125 | |||
Y13936.1 | - | 7 | 1149 |