Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018072.5 | NP_060542.4 | 3 | 394 | ||
GenBank mRNA | AK095692.1 | - | 3 | 357 | ||
AL599555.1 | - | 3 | 368 | |||
BC062442.1 | - | 3 | 345 | |||
BC150614.1 | - | 2 | 264 | 9056984 | ||
DB067322.1 | - | 3 | 394 |