Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCBTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018191.3 | NP_060661.3 | 3 | 369 | ||
XM_005266441.2 | XP_005266498.1 | 2 | 289 | |||
XM_011535133.1 | XP_011533435.1 | 3 | 395 | |||
XM_011535134.1 | XP_011533436.1 | 3 | 295 | |||
XM_011535135.2 | XP_011533437.1 | 2 | 204 | |||
XM_011535136.2 | XP_011533438.1 | 3 | 373 | |||
GenBank mRNA | AF334406.1 | - | 3 | 343 | ||
AJ319660.1 | - | 3 | 310 | |||
AK096654.1 | - | 3 | 369 | |||
AL833821.1 | - | 3 | 279 | |||
BC038104.2 | - | 3 | 363 | 5269720 |