Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018216.2 | NP_060686.2 | 3, 4 | 465 | ||
XM_011541708.2 | XP_011540010.2 | 3, 4 | 431 | |||
XM_017001633.1 | XP_016857122.1 | 3, 4 | 878 | |||
GenBank mRNA | AK001644.1 | - | 3, 4 | 433 | ||
AK223546.1 | - | 3, 4 | 434 | |||
AK295301.1 | - | 3, 4 | 465 | |||
AK309860.1 | - | 2, 3 | 976 | |||
AL442072.1 | - | 3, 4 | 410 | |||
BC043496.1 | - | 3, 4 | 428 | 5244472 | ||
DC316156.1 | - | 3, 4 | 446 | |||
DC327227.1 | - | 3, 4 | 465 |