Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136036.2 | NP_001129508.1 | 2, 3 | 604 | ||
NM_001193328.1 | NP_001180257.1 | 2, 3 | 291 | |||
NM_017865.3 | NP_060335.2 | 2, 3 | 388 | |||
XM_011544220.1 | XP_011542522.1 | 2, 3 | 706 | |||
XM_011544221.1 | XP_011542523.1 | 2, 3 | 706 | |||
XM_011544222.1 | XP_011542524.1 | 2, 3 | 260 | |||
XM_011544223.2 | XP_011542525.1 | 2, 3 | 706 | |||
XM_017001698.1 | XP_016857187.1 | 2, 3 | 260 | |||
XM_017001699.1 | XP_016857188.1 | 2, 3 | 706 | |||
XM_017001700.1 | XP_016857189.1 | 2, 3 | 260 | |||
XM_017001701.1 | XP_016857190.1 | 2, 3 | 375 | |||
XM_017001702.1 | XP_016857191.1 | 2, 3 | 375 | |||
GenBank mRNA | AK000538.1 | - | 2, 3 | 260 | ||
AK001070.1 | - | 2, 3 | 320 | |||
AK225535.1 | - | 2, 3 | 260 | |||
AK302187.1 | - | 2, 3 | 538 | |||
BC002948.1 | - | 2, 3 | 344 | 3533691 |