Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164391.1 | NP_001157863.1 | 12 | 1243 | ||
NM_001164393.1 | NP_001157865.1 | 9 | 1042 | |||
NM_007358.3 | NP_031384.1 | 11 | 1323 | |||
XM_011541016.1 | XP_011539318.1 | 11 | 1279 | |||
XM_011541019.1 | XP_011539321.1 | 11 | 1071 | |||
XM_017000676.1 | XP_016856165.1 | 11 | 1178 | |||
XM_017000679.1 | XP_016856168.1 | 10 | 1155 | |||
GenBank mRNA | AF072814.1 | - | 10 | 1021 | ||
AF073293.1 | - | 1 | 525 | |||
AF086908.1 | - | 10 | 1124 | |||
AJ010014.1 | - | 11 | 1274 | |||
AK290318.1 | - | 11 | 1275 | |||
AK302776.1 | - | 12 | 1182 | |||
AK302900.1 | - | 9 | 994 | |||
BC001897.1 | - | 11 | 1242 |