Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003692.1 | NP_001003692.1 | 5, 6 | 521 | ||
NM_001318129.1 | NP_001305058.1 | 5, 6 | 475 | |||
NM_019103.2 | NP_061976.1 | 6, 7 | 636 | |||
GenBank mRNA | AK292671.1 | - | 5, 6 | 502 | ||
AL160311.1 | - | 5, 6 | 520 | |||
BC009717.1 | - | 5, 6 | 486 | 3890730 | ||
BQ931527.1 | - | 5, 6 | 449 | |||
CR456353.1 | - | 4, 5 | 408 | |||
CR457393.1 | - | 4, 5 | 385 |