Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199699.1 | NP_001186628.1 | 5 | 1094 | ||
NM_020162.3 | NP_064547.2 | 6 | 1255 | |||
GenBank mRNA | AK025625.1 | - | 6 | 1056 | ||
AK295074.1 | - | 5 | 863 | |||
AK295760.1 | - | 6 | 1255 | |||
AK314580.1 | - | 6 | 1223 | |||
AL359945.1 | - | 2 | 233 | |||
BC030017.1 | - | 5 | 781 | |||
BC042040.1 | - | 3 | 626 | |||
CR936655.1 | - | 2 | 233 |