Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001279349.1 | NP_001266278.1 | 3 | 723 | ||
NM_001279350.1 | NP_001266279.1 | 5 | 1061 | |||
NM_001279351.1 | NP_001266280.1 | 5 | 922 | |||
NM_020385.3 | NP_065118.2 | 5 | 1239 | |||
NR_103995.1 | 3 | 698 | ||||
NR_103996.1 | 4 | 892 | ||||
GenBank mRNA | AF273304.1 | - | 5 | 1187 | ||
AF295774.1 | - | 5 | 1187 | |||
AK000247.1 | - | 2 | 100 | |||
AK025493.1 | - | 5 | 1093 | |||
AK295982.1 | - | 5 | 922 | |||
AK304547.1 | - | 5 | 977 | |||
AK310164.1 | - | 3 | 601 | |||
AK314333.1 | - | 5 | 1153 | |||
AK315904.1 | - | 6 | 912 | |||
AL136894.1 | - | 5 | 1170 | |||
BC009274.2 | - | 5 | 1142 | 3958331 | ||
BC071691.1 | - | 4 | 751 | |||
BX333016.2 | - | 3 | 613 |