Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAOK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020791.2 | NP_065842.1 | 13 | 1429 | ||
NM_025142.1 | NP_079418.1 | 13 | 1429 | |||
XM_011525060.2 | XP_011523362.1 | 13 | 1451 | |||
GenBank mRNA | AB037782.1 | - | 13 | 1387 | ||
AF263312.1 | - | 12 | 1235 | |||
AK024376.1 | - | 7 | 1084 | |||
AY049015.1 | - | 12 | 1319 | |||
BC133039.1 | - | 13 | 1425 | 40147061 | ||
BC144067.1 | - | 13 | 1424 | 9052587 |