Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282660.1 | NP_001269589.1 | 10, 11 | 2435 | ||
NM_001282661.1 | NP_001269590.1 | 12, 13 | 2697 | |||
NM_001282662.1 | NP_001269591.1 | 13, 14 | 2847 | |||
NM_001318139.1 | NP_001305068.1 | 11, 12 | 2572 | |||
NM_020831.4 | NP_065882.1 | 13, 14 | 2847 | |||
XM_005261692.1 | XP_005261749.1 | 12, 13 | 2652 | |||
XM_005261694.1 | XP_005261751.1 | 12, 13 | 2742 | |||
XM_011530283.1 | XP_011528585.1 | 14, 15 | 2955 | |||
XM_011530285.1 | XP_011528587.1 | 14, 15 | 2866 | |||
XM_011530287.1 | XP_011528589.1 | 6, 7 | 1834 | |||
XM_017028888.1 | XP_016884377.1 | 13, 14 | 2930 | |||
XM_017028889.1 | XP_016884378.1 | 9, 10 | 3930 | |||
GenBank mRNA | AB037859.2 | - | 13, 14 | 2819 | ||
AF368061.1 | - | 13, 14 | 2772 | |||
AJ297257.1 | - | 10, 11 | 2429 | |||
AJ297258.1 | - | 13, 14 | 2829 | |||
AL713710.1 | - | 5, 6 | 1642 | |||
BC115039.1 | - | 13, 14 | 2813 | 5551767 | ||
BC139730.1 | - | 11, 12 | 1600 | |||
CR456522.1 | - | 10, 11 | 2246 | |||
HG764171.1 | - | 11, 12 | 2572 |