Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128932.1 | NP_001122404.1 | 9 | 1097 | ||
NM_021187.3 | NP_067010.3 | 8 | 1519 | |||
GenBank mRNA | AF236085.1 | - | 8 | 1169 | ||
AK125714.1 | - | 5 | 1224 | |||
AK289424.1 | - | 8 | 1087 | |||
AL833940.1 | - | 8 | 1069 | |||
BC016853.1 | - | 9 | 1097 | 3846027 |