Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021205.5 | NP_067028.1 | 3 | 1188 | ||
NR_037962.1 | 3 | 646 | ||||
GenBank mRNA | AB051826.1 | - | 3 | 601 | ||
AB074878.1 | - | 3 | 661 | |||
AB209511.1 | - | 4 | 720 | |||
AF211836.1 | - | 3 | 1144 | |||
AF251701.1 | - | 3 | 381 | |||
AF282258.1 | - | 3 | 761 | |||
AF378087.1 | - | 3 | 522 | |||
AK289971.1 | - | 3 | 774 | |||
BC040076.1 | - | 3 | 769 | 5768957 |