Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PZP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002864.2 | NP_002855.2 | 10 | 1039 | ||
XM_011520804.2 | XP_011519106.1 | 10 | 1038 | |||
XM_011520805.2 | XP_011519107.1 | 10 | 1038 | |||
XM_011520806.1 | XP_011519108.1 | 10 | 1038 | |||
XM_011520807.2 | XP_011519109.1 | 10 | 1038 | |||
XM_017019763.1 | XP_016875252.1 | 10 | 1038 | |||
XM_017019764.1 | XP_016875253.1 | 10 | 1038 | |||
XM_017019765.1 | XP_016875254.1 | 10 | 1038 | |||
XM_017019766.1 | XP_016875255.1 | 9 | 1187 | |||
XM_017019767.1 | XP_016875256.1 | 10 | 1038 | |||
GenBank mRNA | AK313639.1 | - | 10 | 1038 | ||
BC111756.1 | - | 9 | 1187 | 40027384 | ||
X54380.1 | - | 10 | 1039 |