Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199839.1 | NP_001186768.1 | 3, 4 | 626 | ||
NM_004050.4 | NP_004041.1 | 3, 4 | 671 | |||
GenBank mRNA | AK289519.1 | - | 3, 4 | 626 | ||
BC021198.2 | - | 2, 3 | 608 | 3944307 | ||
BC104789.1 | - | 3, 4 | 607 | 8143792 | ||
BC113522.1 | - | 3, 4 | 607 | 8322574 | ||
BT019549.1 | - | 1, 2 | 442 | |||
D87461.1 | - | 3, 4 | 618 | |||
U59747.1 | - | 1, 2 | 442 |