Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126128.1 | NP_001119600.1 | 4 | 1494 | ||
NM_021935.3 | NP_068754.1 | 3 | 1431 | |||
XM_017006974.1 | XP_016862463.1 | 4 | 1428 | |||
GenBank mRNA | AF333025.1 | - | 3 | 1288 |