Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPN1SW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001708.2 | NP_001699.1 | 4 | 820 | ||
GenBank mRNA | DQ822478.1 | - | 4 | 820 |
Set Membership: |
G-protein coupled receptor |